Possible cause | Solution |
---|---|
Cell not properly installed | Verify that the cell front cover is flush against the component panel. If necessary, tighten the two mounting screws. When the cell is correctly installed and the screws tightened, an electronics connector on the cell plugs into a receptacle inside the Dionex Inuvion IC System. |
No flow from pump | This condition has several possible causes (see Pump does not start and No flow). |