Possible cause | Solution |
---|---|
Incorrect or contaminated eluent | Remake the eluent, using concentrated eluent and ASTM filtered, Type I (18.2 megohm-cm) deionized water that meets the specifications listed in Deionized water requirements for IC. |
Contaminated or degraded sample | Take appropriate precautions when preparing and storing samples to prevent contamination and degradation (see Preparing samples). |
Contaminated column | Procedure
|