Possible cause | Solution |
---|---|
Column overloading | Dilute the sample (see Dilute samples). |
Liquid leaks | Locate and eliminate any leaks (see Liquid Leaks). |
Incomplete or imprecise filling of sample loop | Procedure
|
Pump not primed properly | Prime the pump (see Prime the analytical pump). |