“Ghosting” is the appearance of extraneous peaks in a chromatogram. These may be late-eluting peaks from a previous injection, or they may be the result of an operating issue (for example, contaminated eluent or a malfunctioning valve). These peaks may co-elute with peaks of interest, resulting in nonreproducible peak heights and/or areas.