Possible cause | Solution |
---|---|
Restriction in the system plumbing | Procedure
|
Plugged or damaged fitting | To isolate the faulty fitting, loosen each fitting until the pressure returns to normal. Repair or replace the fitting (see Replace tubing and fittings). |
Flow rate through the columns too high | Set the correct flow rate for your application. |
Clogged column bed supports | Refer to the instructions in the column manual for troubleshooting guidance. |
Contaminated columns | Clean the columns as instructed in the column manual or replace the guard column. |
Plugged injection valve or auxiliary valve passages | Rebuild the valve (see Rebuild the injection valve or auxiliary valve). |