Possible cause | Solution |
---|---|
No power (front Power LED indicator unlit) | Procedure
|
No communication between Dionex Inuvion IC System and Chromeleon CDS | Verify that the USB cable is connected correctly. |
Pump is turned off | Turn on the pump from the Inuvion ePanel. |
Flow rate is set to 0 | Set the flow rate from the Inuvion ePanel. |