Excessive pressure fluctuations (more than 3% difference from one pressure reading to the next) indicate that the pump is out of prime.
Possible cause | Solution |
---|---|
Empty eluent bottle and/or no eluent connected | Fill the bottle. Verify that all connections are secure and then tighten as required. |
Eluent improperly or insufficiently degassed | Procedure
|
End-line filter is dirty or clogged | New end-line filters (P/N 045987) are pure white. Procedure
|
Blockages in inlet tubing | Check for kinked or clogged tubing, which may be causing the pump to be “starved” for eluent. If necessary, replace the tubing and fittings (see Replace tubing and fittings). |
Dirty pump check valve | Clean or replace the pump check valve (see Replace the pump check valves). |
Liquid leaks at junction between pump head and pump casing | Procedure
|
Liquid leaks from seal wash port | Replace the piston seal (see Remove and replace the pistons, analytical pump seals, and O-rings). |
Scratched pump piston | Inspect the pump pistons. If a piston is scratched, replace it (see Remove and replace the pistons, analytical pump seals, and O-rings). |