Possible cause | Solution |
|---|---|
Leaking fitting | Locate the source of the leak. Tighten any tubing fittings or connections and replace them as needed (see Replace tubing and fittings). |
Broken liquid line | When replacing tubing and fittings, the replacements must be of the same length and internal diameter (see Replace tubing and fittings). |
Blocked or improperly installed line | Make sure that no tubing is crimped or blocked. For blocked waste tubing, verify that it is not elevated at any point after it exits the Dionex Inuvion IC System. Waste tubing lines use gravity to drain into the waste container. Replace any blocked tubing. For more information, refer to the suppressor manual. |
Loose pump check valve | Procedure
|
Leaking seal wash port | Leaking from the seal wash port indicates a leaking piston seal. Replace the seal (see Remove and replace the pistons, analytical pump seals, and O-rings). |
Damaged pump piston seal | Procedure
|
Pump head not tight against casing | Carefully tighten the pump head mounting nuts just until the leak stops. Do not overtighten! |
Leaking pressure transducer | Procedure
|
Leaking pump head priming valve | Procedure
|
Leaking suppressor | Refer to the suppressor manual for troubleshooting procedures. |
Leaking injection valve or auxiliary valve | Procedure
|
Leaking detector cell | Procedure
|