The following table lists the most frequently observed Dionex Inuvion IC System error messages, their default severity levels, and troubleshooting steps.
Default severity | Error message | Troubleshooting steps |
---|---|---|
Ambient temperature sensor malfunction | The temperature sensor on the CPU board has failed. Procedure
| |
The auxiliary valve is disconnected | The auxiliary valve is disconnected from the component panel. Procedure
| |
CD board hardware revision does not match the FPGA code version | The conductivity detector board installed in the system is not compatible with the code for the FPGA (field-programmable gate array). Procedure
| |
Cell heater exceeds safe temperature limit | The temperature of the cell heater exceeds the maximum allowed. This may occur, for example, if the system is operating in an environment in which the temperature is greater than 70 °C (158 °F). Procedure
| |
Cell temperature is lower than expected | Cell temperature is less than 2 °C (35.6 °F). Procedure
| |
Column heater exceeds safe temperature limit | The temperature of the column heater exceeds the maximum allowed. This may occur, for example, if the system is operating in an environment in which the temperature is greater than 70 °C (158 °F). Procedure
| |
The column heater is disconnected | Usually indicates that the column heater is unplugged from the component panel. Procedure
| |
Column heater memory reset to defaults | Occurs when no valid data is read from the column heater memory after system power-up. As a result, all parameters are reset to their defaults. Procedure
| |
Column heater open circuit | Usually indicates that the column heater is unplugged from the component panel. Procedure
| |
Column heater short circuit | Indicates a short circuit of the thermistor input that is used to measure the column heater temperature. Procedure
| |
Column heater temperature calibration failed | Occurs when the calibration value for the column heater temperature is out of range. This causes the calibration procedure to fail. Procedure
| |
Compression limit reached at 100% | Occurs when the pump is unable to regulate the pressure any further (i.e., the Maintenance_Pump_K_Value cannot be increased). It may also indicate an overcurrent in the pump drive bridge. Procedure
| |
The conductivity detector is disconnected | Error indicates that the conductivity detector is disconnected from the component panel. Procedure
| |
CR-TC board hardware revision does not match the FPGA code version | Occurs when the system receives a Dionex CR-TC command from Chromeleon CDS but is unable to recognize the Dionex CR-TC controller board. Procedure
| |
CR-TC memory reset to defaults | Occurs when there is a Dionex CR-TC memory read/write failure. As a result, all parameters are reset to their default values. Procedure
| |
CR-TC not installed | Occurs when a command is issued to control a Dionex CR-TC that has not been installed (or when a Dionex CR-TC is installed but has not yet been configured). Procedure
| |
CR-TC over current—hardware detected | Occurs when the current applied to the Dionex CR-TC exceeds the maximum current (120 mA) allowed. (The current will be automatically turned off to prevent damage to the Dionex CR-TC.) Procedure
| |
CR-TC over power | Occurs when, to maintain the selected current, the system must apply a higher voltage than the Dionex CR-TC can support. Procedure
| |
CR-TC over voltage—firmware detected CR-TC over voltage—hardware detected | Occurs when the Dionex CR-TC voltage exceeds the commanded voltage from the user. Procedure
| |
CR-TC short circuit | Occurs when the current applied to the Dionex CR-TC exceeds the maximum value allowed (250 mA). The Dionex CR-TC is automatically turned off to prevent damage. Procedure
| |
CR-TC status not ready | Occurs when the FPGA (field-programmable gate array) attempts to access the Dionex CR-TC EEPROM or consumable monitoring (1-wire) chip while the Dionex CR-TC is still busy with a previous command. Procedure
| |
CR-TC stops because EGC is off | Occurs if you turn off the pump flow while the Dionex EGC current (and Dionex CR-TC) are on. When the pump flow rate is 0.00 mL/min, the Dionex EGC is automatically turned off. When the Dionex EGC is off, the Dionex CR-TC is automatically turned off to prevent damage. Procedure
| |
CR-TC stops for zero flow | Occurs when the pump stops unexpectedly. Procedure
| |
Degasser is in Fault status | Indicates a problem in the Dionex Inuvion RFIC Degasser electronics. Procedure
| |
Detector memory reset to defaults | Occurs when there is a conductivity detector memory read/write failure. As a result, all parameters are reset to their default values. Procedure
| |
The device is busy—please retry after 1 minute | Occurs if you attempt to send a pump command while the pump is still busy with a previous action (for example, docking or undocking). Procedure
| |
EG board hardware revision does not match the FPGA code version | Indicates that the eluent generator board installed in the system is not compatible with the code for the FPGA (field-programmable gate array). Procedure
| |
EG status not ready | Occurs when the eluent generator is not plugged in, or when the installed device is the wrong product type. Procedure
| |
EGC cartridge disconnected | Occurs when the Dionex EGC is disconnected from the eluent generator electronics. Procedure
| |
EGC cartridge not installed | Occurs when a command is issued to control a Dionex EGC that has not been installed yet (or has been installed, but not yet configured in Chromeleon CDS). Procedure
| |
EGC invalid activation date | Occurs if the activation date for the Dionex EGC is invalid. This may indicate a problem with the memory chip in the Dionex EGC. Procedure
| |
EGC invalid concentration | Occurs when the eluent concentration is outside the range allowed by the Dionex EGC. Procedure
| |
EGC invalid ion count | Occurs when the ion count reported by the Dionex EGC is invalid. This may indicate a problem with the memory chip in the Dionex EGC. Procedure
| |
EGC invalid serial number | Occurs when the serial number reported by the Dionex EGC is invalid. This may indicate a problem with the memory chip in the Dionex EGC. Procedure
| |
EGC memory reset to defaults | Occurs when no valid data can be read from Dionex EGC memory after system power-up. As a result, all parameters are reset to their defaults. Procedure
| |
EGC over current—firmware detected EGC over current—hardware detected | Occurs when the current applied to the Dionex EGC exceeds the maximum allowable current. Under these conditions, the Dionex EGC is turned off automatically to prevent damage. This error may also occur if liquid flow to the cartridge is interrupted or blocked. Procedure
| |
EGC over power | Occurs when, in order to maintain the selected current, the power supply must supply a higher voltage than the Dionex EGC can support. Procedure
| |
EGC over voltage—firmware detected EGC over voltage—hardware detected | Occurs when the voltage applied to the Dionex EGC exceeds the maximum recommended level. (Under this condition, the Dionex EGC is turned off automatically to prevent damage.) This error may also occur when liquid flow to the Dionex EGC is interrupted. Procedure
| |
EGC stops for zero flow | Occurs when the pump flow stops. Procedure
| |
EGC wrong cartridge error | Occurs when a Dionex EGC that is not supported by the Dionex Inuvion IC System is plugged in. Procedure
| |
The eluent bottle is empty | Occurs if there is no eluent in the eluent bottle. Procedure
| |
The eluent generator cartridge has expired. Will abort the sequence after current injection. | Indicates that the Dionex EGC is expended. (The Dionex EGC expiration date is 2 years from the date of manufacture.) Chromeleon CDS will complete the current injection and then abort the sequence. Procedure
| |
Eluent generator cartridge ion count is low | Occurs when the Dionex EGC ion count is nearing depletion. Procedure
| |
The eluent generator cartridge’s expiration date is approaching | Occurs when the Dionex EGC is nearing its expiration date. (The Dionex EGC expiration date is 2 years from the date of manufacture.) Although it is possible to continue operation with the Dionex EGC after the expiration date, performance may be impaired until a new cartridge is installed. Procedure
| |
Entry fan tacho timeout | The entry fan on the rear panel of the Dionex Inuvion IC System improves air convection inside the system. This error occurs when the tachometer that measures the speed of the entry fan times out. This indicates that either the fan is stalled or the fan motor drive is faulty. Procedure
| |
Flash memory reset to defaults | Indicates a malfunction in the system electronics. Procedure
| |
Flow is on—cannot undock piston | Occurs if you select the Pistons_Undock check box in the Chromeleon CDS Command window while the analytical pump is running. Procedure
| |
FPGA flash memory erase failed | Occurs when an erase operation fails to delete all data from the FPGA (field-programmable gate array) flash memory. Procedure
| |
FPGA flash memory write failed | Occurs when a write operation fails to copy all data to the FPGA (field-programmable gate array) flash memory. Procedure
| |
FPGA Main is not configured | Occurs when the FPGA (field-programmable gate array) on the Main board has not been configured yet. When this occurs, the board is unable to accept code. Procedure
| |
The Hall sensor did not signal the zero position over more than 5 revolutions | Occurs if the Hall sensor fails to signal the zero position of the CAM of the pump motor for more than five revolutions. The zero position is the home position of the pump (i.e., the reference point as it rotates around a fixed axis). Procedure
| |
The Hall sensor reported an impossible zero position | Occurs if the Hall sensor reports a mismatch between the pump motor rotational counts and the CAM rotations. The zero position is the home position of the pump (i.e., the reference point as it rotates around a fixed axis). This may indicate a calibration error in the CAM position or physical damage to the encoder on the pump mechanism. Procedure
| |
High pressure valve busy | Occurs when the high-pressure valve is unable to complete the required action. Procedure
| |
High pressure valve failed to reach commanded position | Occurs when the high-pressure valve fails to switch position within 1 second of being toggled. Procedure
| |
Inject valve busy | Occurs when the injection valve is unable to complete the required action. Procedure
| |
Inject valve failed to reach commanded position | If the injection valve fails to switch position within 1 second of being toggled, the instrument control firmware reports an error to Chromeleon CDS and this error message appears. Procedure
| |
The inject valve is disconnected | Occurs when the injection valve cannot be detected. Procedure
| |
The leak sensor is wet | The leak sensor is installed in the drip tray located near the bottom of the component panel. This error occurs when liquid accumulates in the drip tray. Procedure
| |
The low pressure valve 1 is disconnected The low pressure valve 2 is disconnected | Occurs when the low-pressure valve cannot be detected. Procedure
| |
Motor power failure detected | This error occurs when the pump motor fails. This typically occurs when the system power is turned off while the pump is running. You may also notice a significant pressure transducer offset. Procedure
| |
Pressure exceeds upper limit | If the system pressure exceeds the set limit, this error occurs and the pump stops. Procedure
| |
Pressure fallen below lower limit | If the system pressure falls below the set limit, this error occurs and the pump stops. Pressure limits can be set in the Chromeleon Instrument Configuration Manager or in the instrument method. Procedure
| |
The pressure sensor is malfunctioning | Occurs when the pressure sensor fails to detect a rise in pressure while the pump is turned on. This will cause the pump to shut down. The error may also occur if the pressure remains at a fixed value while the pump is off. Procedure
| |
Pressure slope calibration error | Occurs when the user attempts to calibrate the pressure slope when the pressure is less than 3 MPa (500 psi). Procedure
| |
Pressure was larger than 725 psi in maintenance mode | Occurs if you undock the pump head (to perform maintenance) when the purge valve is closed. Procedure
| |
Preventive maintenance interval is approaching | Occurs 21 days before the 1-year anniversary of the last annual preventive maintenance procedure for the Dionex Inuvion IC System. Procedure
| |
Pump board memory reset to defaults | Occurs when there is a diagnostic failure. Procedure
| |
Pump flow limit exceeded | Occurs when you attempt to select a flow rate that is outside the flow rate range. Procedure
| |
Pump FPGA hardware over pressure. The pressure limiter circuit detected an overpressure condition. | Indicates a problem in the pump controller electronics. Procedure
| |
Pump FPGA motor encoder error | Indicates a problem in the pump controller electronics. Procedure
| |
The pump is not ready because it was switched into standby mode | Occurs when the pump is off while in standby mode. Procedure
| |
The pump is not ready because the self-test did not finish successfully | Occurs when the pump self-test is not able to finish. During the self-test, which starts automatically each time the system is powered up, the pump moves forward and backward slightly to verify that it works properly. Procedure
| |
Pump is still in undock position | Occurs if you try to turn on the pump while the piston is undocked. Procedure
| |
Pump motor over current | Indicates a problem in the pump controller electronics. Procedure
| |
Pump motor position error. The motor control was not able to move the motor to the correct position. | Indicates a problem in the pump controller electronics. Procedure
| |
Pump Off for lost USB communication | Occurs when the Dionex Inuvion IC System is unable to communicate with Chromeleon CDS. Procedure
| |
Pump pressure hardware error | Indicates a problem in the pump controller electronics. Procedure
| |
Pump pressure sensor calibration failed | Indicates a problem in the pump controller electronics. Procedure
| |
Pump priming cannot start—system pressure too high | Occurs if the user attempts to prime the pump while the priming valve is closed. Procedure
| |
Pump seal wash stopped because it is switched to standby | Occurs when you try to turn on the seal wash pump while the piston is undocked. Procedure
| |
Pump stopped due to lost USB communication error | Occurs when the Dionex Inuvion IC System is unable to communicate with Chromeleon CDS. Procedure
| |
Purge pressure limit exceeded | Occurs when you try to purge the pump when the purge knob is closed or when there is a blockage in the purge waste tubing. This error may also indicate a problem in the pump controller electronics. Procedure
| |
Suppressor board hardware revision does not match the FPGA code version | Indicates that the suppressor board installed in the system is not compatible with the code for the FPGA (field-programmable gate array). Procedure
| |
Suppressor memory reset to defaults | Occurs when the suppressor type is invalid, or when the local calibration memory is blank. Procedure
| |
Suppressor not installed | Occurs when the suppressor is on, but no suppressor is detected. Procedure
| |
Suppressor over current—firmware detected Suppressor over current—hardware detected | Indicates that the suppressor is depleted or dirty, or that there is a malfunction in the suppressor controller electronics. Procedure
| |
Suppressor over power | Occurs when, to maintain the selected current, the Dionex Inuvion IC System is required to apply a higher voltage than the suppressor can support. Procedure
| |
Suppressor over voltage—firmware detected Suppressor over voltage—hardware detected | Occurs if you turn on the system and it is unable to establish a connection with the suppressor. Procedure
| |
Suppressor status not ready | Occurs if no suppressor is installed, or if the installed suppressor is the wrong type. Procedure
| |
Suppressor stops for zero flow | Appears after selecting the Shutdown button on the Inuvion ePanel. It also appears if the pump flow stops unexpectedly while the suppressor is on. To prevent suppressor damage, the suppressor automatically turns off when there is no flow. Procedure
| |
Suppressor type has changed | Occurs when the selected suppressor type is not appropriate for the Dionex EGC. Procedure
| |
There is not enough pressure data | Occurs when there is an internal communication error. Procedure
| |
The waste bottle is full | Occurs when the waste container is full. Procedure
| |
Wrong hardware configuration | Occurs when there is a mismatch between the configuration in Chromeleon CDS and the hardware that is installed. Procedure
|