Procedure
  1. Turn off the Dionex Inuvion IC System main power switch on the rear panel.
  2. Main power switch and fuse holder
    Figure Main power switch and fuse holder
  3. DANGER
    HIGH VOLTAGE
    Disconnect the main power cord from its source and also from the rear panel of the Dionex Inuvion IC System.
  4. The fuse holder is part of the main power receptacle on the Dionex Inuvion IC System rear panel (see Change the main power fuses). To remove the fuse holder, squeeze the tab on top of the holder to release it and pull the holder straight out of its compartment.
  5. Main power fuse holder
    Figure Main power fuse holder
  6. Replace the two fuses in the holder with new fuses (P/N 00006-03-00013) rated 8 amps. Thermo Fisher Scientific recommends always replacing both fuses.
  7. Reinsert the fuse holder into its compartment and push it into the instrument until the tab locks in place.
  8. Reconnect the main power cord and turn on the power.