Procedure
  1. Make sure the eluent bottle is filled.
  2. Make sure the ELUENT BOTTLE OUT line from the bottle cap is connected to the ELUENT IN line, which extends from the tubing chase on the top cover of the Dionex Inuvion IC System.
  3. Make sure the Dionex EGC electrical cable is connected to the EGC connector.
  4. Make sure the Dionex CR-TC electrical cable is connected to the CR-TC/EPM connector.