Procedure
- Press the main power switch on the Dionex Inuvion IC System rear panel.
- Turn on the power of the computer and the autosampler (if installed).
Feature | Power-up condition |
---|---|
Pump | Off |
Injection valve | Load position |
Cell | Reading current value |
Suppressor | Off1) |
Cell heater | Off. When turned on, the cell returns to the last value used. When the system is turned on for the first time, the default is 35°C. |
Column oven temperature (optional) | Off. When turned on, the column oven returns to the last value used. When the system is turned on for the first time, the default is 30°C. |
Eluent generator | Off1) |
Dionex CR-TC | Off |