Install the digital pressure regulator - Dionex Inuvion - Dionex Inuvion Operating Manual - User maintenance - Install the digital pressure regulator - Install and pressurize the digital gas pressure regulator - Dionex Inuvion
Chromatography > Ion Chromatography > Dionex Inuvion
Document Type
User Guide
Operating Manual
Procedure
Unpack the components from the Digital Pressure Regulator Kit and place them on a clean surface near the Dionex Inuvion IC System.
Turn off the Dionex Inuvion IC System main power switch on the rear panel of the system.
Locate the digital display blank cover just above the conductivity detector. Remove the two screws on the cover plate and save the screws for reinstallation.
Figure Blank cover without digital pressure regulator display
Loosen the twist tie securing the cable to the cover plate.
Figure Digital display back cable
Connect the digital display cable to the digital display connector.
Figure Digital pressure regulator cable connection
Place the digital regulator display into position. Fasten both screws to secure the display to the assembly.
Figure Digital regulator display view
On the back of the Dionex Inuvion IC System, remove the five 3 mm hex bolts securing the blank cover.
Figure Digital pressure regulator port location (with port cover)
Untie the twist tie securing the inside cable to the back of the cover plate.
Connect this back cable to the digital pressure regulator connector.
Figure Digital regulator cable connection
Put the digital pressure regulator assembly into position and fasten all five hex bolts.