Install or replace the column heater - Install or replace the column heater - Dionex Inuvion Operating Manual - Dionex Inuvion - Dionex Inuvion Operating Manual - User maintenance - Install or replace the column heater - Dionex Inuvion - topic
Dionex Inuvion Operating Manual
- Product Tree
- Chromatography > Ion Chromatography > Dionex Inuvion
- Document Type
- User Guide
- Operating Manual
- Locate the column heater kit (P/N 22185-62400) and a medium-sized flathead screwdriver.
- Open the front door of the system and locate the column mounting plate at the right side of the system.
- Remove the magnetic column heater cover by pulling it straight out.
- Unscrew the three screws for the column mounting plate and remove it by pulling it straight out.
- Align the column heater with the four pegs and electrical connector on the system component panel; then push the column heater into place.
- Use the thumbscrews to fasten the column heater to the mounting plate. You will need to use a screwdriver for the top screw, due to limited finger access.
- Install the columns in the new heater by gently snapping them into the clips, and then snap the magnetic cover back into place.
- Close the system door and turn on the main power switch.