Procedure
  1. From the Chromeleon CDS ePanel, turn off the analytical pump.
  2. Disconnect the Dionex EGC electrical cable from the EGC connector on the Dionex Inuvion IC System top cover (see the following figure).
  3. Remove the Dionex EGC VENT line from the cartridge by unscrewing the luer lock from the luer adapter.
  4. TIP
    If necessary, use a wrench to hold the luer adapter in place while unscrewing the vent line.
  5. Install the luer cap (P/N 053981) provided with the Dionex EGC onto the luer adapter. This prevents leaks from the vent opening when you turn over the cartridge (see Step 6).
  6. Lift the Dionex EGC straight up and out of the cartridge holder.
  7. Turn the Dionex EGC upside down and set it in the cartridge service area on top of the Dionex Inuvion IC System.
  8. Dionex EGC service area within the eluent organizer area
    Figure Dionex EGC service area within the eluent organizer area
  9. Disconnect the EGC IN and EGC OUT lines from the INLET and OUTLET ports on the Dionex EGC.
  10. Follow the instructions in the Dionex EGC manual to prepare an expended cartridge for disposal or to store a partially used cartridge for later use.