To use an autosampler, the Dionex Inuvion IC System injection valve must be specified as the injection valve and linked to the autosampler in the Chromeleon CDS instrument. For more information, refer to the Chromeleon CDS Help or the operator’s manual for the autosampler.