There are two techniques for loading samples into the sample loop:
  • With an autosampler
  • With a syringe or vacuum syringe via the injection port on the Dionex Inuvion IC System front panel
For autosampler injections, the injection port tubing must be disconnected from the Dionex Inuvion IC System injection valve and replaced by the autosampler outlet tubing. Other setup requirements vary, depending on the autosampler model.
Samples can be injected using either the standard injection valve or the optional auxiliary valve. The injection valve (or auxiliary valve) must be specified as the injection valve and linked to the autosampler in the Chromeleon CDS for the configured instrument. For more information, refer to the Chromeleon CDS Help.