The following overview details simplified steps for routine operation of the Dionex Inuvion IC System.
Figure System operation overview
Check all connections Power up the system Prime the pump Set operating conditions Equilibrate the system Prepare samples Verify operating status Process samples Start Chromeleon CDS Set up the eluent reservoir (or RFIC components)