This section describes initial setup and routine operating procedures for the Dionex Inuvion IC System.
Chromeleon CDS automatically detects and configures Dionex Inuvion IC System components when the system is first started. Instructions in this chapter detail the initial setup (including configuring the system in a Chromeleon CDS instrument) following installation by a Thermo Fisher Scientific service engineer.
Components added after initial setup must be added manually because Chromeleon CDS detects components during initial setup only. Complete resetting of the Dionex Inuvion IC System will cause Chromeleon CDS to automatically detect all currently installed components, but is not generally recommended.
For more information about installation and maintenance procedures that users can perform, see User maintenance.