A low-pressure three-way (or three-port) valve (P/N B51001290) can be used for on/off control of liquid flow (for example, to turn flow on and off from a reagent or external regenerant bottle). Up to two low-pressure valves can be supported.
A low-pressure three-way (or three-port) valve (P/N B51001290) can be used for on/off control of liquid flow (for example, to turn flow on and off from a reagent or external regenerant bottle). Up to two low-pressure valves can be supported.