The USB receptacle provides a connection to the computer on which Chromeleon CDS is installed. Three USB ports are available for connections to other USB devices.
WARNING
Do not exceed USB cable lengths of 10 m (32.8 feet) and do not use additional hubs or extenders. Excessive cable length or unapproved hubs or extenders can increase RF electromagnetic emissions or affect the sensitivity of the Dionex Inuvion IC System.