The analog output connector outputs conductivity data (as a 0 to 1 V signal) to an integrator or recording device.
The analog output connector outputs conductivity data (as a 0 to 1 V signal) to an integrator or recording device.