The following components are accessible from the Dionex Inuvion IC System rear panel:
- Analog output connector
- USB connectors
- TTL and relay connector
- Tubing clips
- Gas regulator mounting location
- Low-pressure valve
- Power switch
- Main power receptacle
The following image shows the rear panel.