Both the analytical and guard columns are packed with resin and perform separation of the sample ions. The main function of the guard column is to trap contaminants and remove particulates that might damage the analytical (or separator) column.
Both the analytical and guard columns are packed with resin and perform separation of the sample ions. The main function of the guard column is to trap contaminants and remove particulates that might damage the analytical (or separator) column.