The conductivity detector consists of a temperature-controlled conductivity cell and electronics that produces conductivity data and relays it to the computer and analog output. The flow-through conductivity cell measures the electrical conductance of analyte ions as they pass through the cell via two passivated 316 stainless steel electrodes which are permanently sealed into the PEEK cell body. The cell design provides efficient sweep-out, low volume, and low dispersion.