A leak sensor is installed in the drip tray at the bottom of the front panel. If liquid accumulates in the tray, the front panel Alarm LED lights up and an error message is logged in the Instrument audit trail.