The suppressor reduces the eluent conductivity and enhances the conductivity response of the sample ions, thereby increasing detection sensitivity. For more information about the suppressors (or guidelines for selecting a suppressor for an application), refer to the suppressor manuals.
The Dionex Inuvion IC System is compatible with the suppressors listed in the following table.
Suppressors for the Dionex Inuvion IC System
Part number
Suppressor
085090
Dionex ACRS 500 Anion Dynamically Regenerated Suppressor (4 mm)
085091
Dionex ACRS 500 Anion Dynamically Regenerated Suppressor (2 mm)
088666
Dionex ADRS 600 Anion Dynamically Regenerated Suppressor (4 mm)
088667
Dionex ADRS 600 Anion Dynamically Regenerated Suppressor (2 mm)
302661
Dionex AERS 500e Electrolytically Regenerated Suppressor (4 mm)
302662
Dionex AERS 500e Electrolytically Regenerated Suppressor (2 mm)
085029
Dionex AERS 500 Carbonate Electrolytically Regenerated Suppressor (4 mm)
085028
Dionex AERS 500 Carbonate Electrolytically Regenerated Suppressor (2 mm)
085092
Dionex CCRS 500 Cation Dynamically Regenerated Suppressor (4 mm)
085093
Dionex CCRS 500 Cation Dynamically Regenerated Suppressor (2 mm)
088668
Dionex CDRS 600 Cation Dynamically Regenerated Suppressor (4 mm)
088670
Dionex CDRS 600 Cation Dynamically Regenerated Suppressor (2 mm)
302663
Dionex CERS 500e Electrolytically Regenerated Suppressor (4 mm)
302664
Dionex CERS 500e Electrolytically Regenerated Suppressor (2 mm)