The Dionex CR-TC is a high-pressure electrolytically-regenerated trap column. The Dionex CR-TC is designed to remove anionic or cationic contaminants in the eluent or deionized water and to reduce drift during gradient separations. The following consumables can be used with the Dionex Inuvion IC System:
  • Thermo Scientific™ Dionex™ Dionex CR-CTC 600 Continuously Regenerated Cation Trap Column (P/N 088663)
  • Dionex CR-ATC 600 Continuously Regenerated Anion Trap Column (P/N 088662)
For more information, refer to the trap column manual.