Flow output from the pressure transducer continues to the pulse damper, located above the pressure transducer on the component panel of the system. The pulse damper reduces minor pressure variations for improved analysis. From there, flow is directed to the injection valve and the remainder of the chromatography system.