The analytical pump is a microprocessor-based eluent delivery system. The pump’s variable speed, dual-piston series design ensures pulse-free pumping for the most demanding applications.
The analytical pump is a microprocessor-based eluent delivery system. The pump’s variable speed, dual-piston series design ensures pulse-free pumping for the most demanding applications.