The eluent valve (see the following image) controls the flow from the eluent bottle. The eluent valve opens automatically when the pump starts and closes when the pump is turned off. This enables you to perform maintenance procedures on pump components with minimal eluent leakage.