Users can maintain and/or replace user-accessible components installed on the component panel behind the Dionex Inuvion IC System front door. Select the preferred options for components in the Properties dialog box.
Procedure
  1. Open the Chromeleon Instrument Configuration Manager.
  2. Double-click the Inuvion icon under the instrument.
  3. In the Properties dialog box, select the Options tab (see the following figure).
  4. Select the preferred options.
Figure Properties dialog box: Pump tab page
Properties dialog box: Pump tab page