The Power LED indicates whether the Dionex Inuvion IC System power is on. Other LEDs indicate the status of various system functions. Active LEDs are green (except when blinking), with the exception of Alarm, which is red when activated. For details, see the following table.
LED label | If on (solid) | Displayed on... |
---|---|---|
Power | Indicates that the system is on | Dionex Inuvion Core, Dionex Inuvion, Dionex Inuvion RFIC |
Alarm | Indicates that an error occurred; the LED remains on until the error is resolved | Dionex Inuvion Core, Dionex Inuvion, Dionex Inuvion RFIC |
Pump | Indicates that the pump is on and operating normally | Dionex Inuvion Core, Dionex Inuvion, Dionex Inuvion RFIC |
SRS | Indicates that the suppressor is on and operating normally | Dionex Inuvion, Dionex Inuvion RFIC |
RFIC | Indicates that the RFIC is on and operating normally | Dionex Inuvion RFIC |