Figure Dionex Inuvion Core IC System component panel (with column holder and regenerant pump installed)
1234567891011
No.DescriptionNo.Description
1
Eluent monitor (optional)
7
Regen pump (optional)
2
Column holder
8
Tubing chase (to eluent organizer)
3
Injection valve
9
Tubing chase (to waste)
4
Leak sensor
10
Conductivity cell
5
Pump
11
Digital pressure regulator (optional)
6
Seal wash pump (optional)
Figure Dionex Inuvion IC System component panel with column heater cover and auxiliary valve installed