The Dionex Inuvion IC System and its optional accessories offer reliable, configurable, and easy operation across diverse applications through these user-friendly features:
- Simplified flow path
- Easy access to system components for user maintenance and upgrades
- Streamlined Chromeleon CDS user interface
- Smaller size, with reduced footprint and greater portability
- Ergonomic, easily accessed eluent generator cartridge (Dionex EGC 500)
- Wide range of user-orderable and installable accessories
- Automated startup and shutdown procedures
- Faster baseline stability via updated electronics
- Upgraded analytical pump with greater reliability and durability
- Improved conductivity detector with conductivity cell
- Automatic detection of one-wire enabled consumables
- Enhanced tubing management with additional clips and holders
- Improved column oven for uniform and faster heating
- Tubing connections with Thermo Scientific™ Dionex™ IC PEEK™ Viper™ fittings for ease of use and improved connections
- Expanded training and user support
The Dionex Inuvion IC System is available with and without RFIC eluent generation. Systems can be upgraded between models and configurations by Thermo Fisher Scientific service engineers.
Required consumables, including analytical columns, must be purchased to perform applications. Other consumables, such as suppressors and guard columns, are required for most applications, with some exceptions.