Procedure
- If the Dionex DRS 600 is observed to be leaking, see Liquid leaks.
- Ensure the Sample Injection Valve and Recycle Valve are operating correctly. Refer to the valve manuals that accompany the valves. Be sure to check the port faces for damage.
- If the neutralization requires excessive recycling, clean the suppressor membranes (see Cleaning the suppressors).
- If cleaning the suppressor membranes does not restore neutralization efficiency, the suppressor may need to be replaced.
If you cannot solve the problem on your own, contact the Thermo Scientific Regional Office nearest you.