See the following table for troubleshooting tips.
Issue | Procedure |
---|---|
Excessive current applied to the Dionex DRS 600 (Legacy Mode), Dionex ERS 500e or Dionex AERS 500 Carbonate | Procedure
NOTE: During a low flow method (stand-by mode), ensure that the current is lowered to the optimal value based on the reduced flow rate and/or eluent concentration. |
Precipitation on the suppressor membrane or screens (calcium, magnesium, and other metals) | Procedure
|
Outgassing or trapped bubbles in the suppressor regenerant chambers | Procedure
|
Excess suppressor temperature | Procedure
|
Large changes to the flow rate or applied current of the suppressor | Procedure
|
Operating the suppressor with power but no eluent or regenerant flow | Operation of the suppressor without flow may irreversibly damage the suppressor, depending on the amount of applied current and duration. Procedure
|
Incorrect current setting during shutdown, stand-by or startup method | Procedure
|