If the baseline drifts steadily upward, increase the current setting by 5-10% to reduce the background conductivity. As the background conductivity decreases, the baseline drift should decrease.
If the baseline drifts steadily upward, increase the current setting by 5-10% to reduce the background conductivity. As the background conductivity decreases, the baseline drift should decrease.