This topic details the configuration process for a new suppressor in CM Configuration.
Procedure
  1. Plug in the new suppressor, then open CM Configuration.
  2. Do one of the following:
  3. Select the Suppressor tab (ICS-6000).
  4. Select the Electrolytics tab (Integrion).
  5. Double-click the suppressor option, then select OK to confirm the new configuration.
  6. Suppressor configuration in ICS-6000 DC Detector
    Suppressor configuration in ICS-6000 DC Detector
  7. Suppressor configuration in Integrion
    Suppressor configuration in Integrion
  8. Close the Instrument Controller.
  9. You are prompted to confirm changes to the configuration.
  10. On the window that appears, select Save.
  11. Save changes to the instrument configuration
    Save changes to the instrument configuration
  12. NOTE
    If you do not save your changes, CM will not recognize the new suppressor.
Installation summary
Installation summary
CAUTION
Only use the AutoSuppression Recycle Mode for eluents and samples without organic solvents or metallic contaminants such as iron.