Flow restriction or blockage from detector cell or backpressure coils should be eliminated first to avoid irreversible damage to the suppressor.
Task | Procedure |
|---|---|
Backflush the detector cell | Procedure
NOTE: Repeat the backflush procedure if the backpressure remains high. If the backpressure of the detector cell doesn’t come below 70 psi after multiple attempts, replace the detector cell. |
Adjust backpressure coil | Procedure
NOTE: Backpressure coils help to prevent gases generated during AutoSuppression from out-gassing by compressing the bubbles. The bubbles can be occasionally trapped in the detector cell causing noise. For example, carbonate eluent is suppressed to carbonic acid, which is CO2 gas in equilibrium with DI water, and CO2 gas can come out of solution if adequate pressure is not applied. Therefore, we highly recommend the addition of 30-40 psi for carbonate eluent or 10-40 psi for other eluents. |
Backflush suppressor with DI water | Procedure
NOTE: Repeat the backflush procedure if the backpressure remains high. If the backpressure of the suppressor doesn’t come below 200 psi after multiple attempts, replace the suppressor or contact your Service Engineer or Technical Support team. |