Long-term storage (more than 7 days) - ERD 500 - ERS 500e - ERS 500 Carbonate - DRS 600 - Long-term storage (more than 7 days) - ERD 500 - ERS 500e - ERS 500 Carbonate - DRS 600
Dionex DRS 600 / ERS 500e / ERS 500 Carbonate Suppressors User Guide
- Product Tree
- Chromatography > Ion Chromatography > Suppressors > ERD 500
- Chromatography > Ion Chromatography > Suppressors > ERS 500e
- Chromatography > Ion Chromatography > Suppressors > ERS 500 Carbonate
- Chromatography > Ion Chromatography > Suppressors > DRS 600
- Document Type
- User Guide
- Connect the eluent and regenerant chambers in series and flush the Dionex DRS 600, Dionex ERS 500e or Dionex AERS 500 Carbonate with deionized water for 10 minutes at 1.0 mL/min (4 mm) or 0.25 mL/min (2 mm).
- Plug all suppressor ELUENT ports and REGEN ports.
- To resume operation, complete the hydration steps as outlined in Hydration and connect the suppressor to the system. Allow the system to equilibrate before starting the analysis.