See the following table for details on pump trap column installation and regeneration.
Task | Description |
|---|---|
Install the Dionex IonPac ATC-HC Trap Column | The Dionex IonPac ATC-HC Trap Column (Item # 059604) is used to trap anions that may be in the carrier solution that could result in high blanks. The Dionex IonPac ATC-HC should be regenerated prior to installation. |
Regenerate the Dionex IonPac ATC-HC Trap Column | Using a pump other than the carrier pump, the Dionex IonPac ATC-HC should be regenerated and rinsed: Procedure
|
Install the Dionex IonPac CTC-1 Trap Column | The Dionex IonPac CTC-1 Trap Column (Item # 040192) is used to trap cations that may be in the carrier solution that could result in high blanks. The Dionex IonPac CTC-1 should be regenerated prior to installation. |
Regenerate the Dionex IonPac CTC-1 Trap Column | Using a pump other than the carrier pump, the Dionex IonPac CTC-1 should be regenerated and rinsed: Procedure
|