The SP10 Auto Neutralizer Ship Kit (Item # 047950) contains all the components needed to install and operate the Dionex DRS 600 in Neutralization Mode with a pressurized water reservoir.
Procedure
- Make the following air line connections:
- Locate the pieces of tinted 1/8” OD plastic tubing (Item # 030089) supplied in the Ship Kit.
- Push the end of a piece of 1/8” OD tubing over the barbed fitting of the regulator. Connect the other end of the tubing to the source of air pressure.
- Push one end of the second piece of 1/8” OD tubing over the other barbed fitting of the regulator. Push the other end of this tubing over the barbed fitting (Item # 030077) in the pressure inlet of the plastic reservoir.
- Make the following liquid line connections:
- Use a coupler (Item # 039056) to connect one end of the 30” tubing assembly (Item # 035727) that comes in the Auto Neutralization Ship Kit to the water reservoir. Connect the other end of this tubing to the REGEN IN port of the Dionex DRS 600.
- Using a coupler (Item # 039056) and a 1/8” OD piece of tubing (Item # 035728) from the Auto Neutralization Ship Kit, connect one end of this line to the REGEN OUT port of the Dionex DRS 600 and then connect the other end of the line to the Gas Separator Waste Tube.
- Fill the reservoir with water. Make sure the O-ring is inside the cap of the reservoir before screwing the cap onto the reservoir. Screw the cap onto the reservoir tightly and place the reservoir near or on top of the instrument in a secondary waste container such as the Dionex EO.