Sample liquid line connections for Dionex DRS 600 in Neutralization Mode - ERD 500 - ERS 500e - ERS 500 Carbonate - DRS 600 - Sample liquid line connections for Dionex DRS 600 in Neutralization Mode - ERD 500 - ERS 500e - ERS 500 Carbonate - DRS 600
Dionex DRS 600 / ERS 500e / ERS 500 Carbonate Suppressors User Guide
- Product Tree
- Chromatography > Ion Chromatography > Suppressors > ERD 500
- Chromatography > Ion Chromatography > Suppressors > ERS 500e
- Chromatography > Ion Chromatography > Suppressors > ERS 500 Carbonate
- Chromatography > Ion Chromatography > Suppressors > DRS 600
- Document Type
- User Guide
- Install the Dionex DRS 600 in the chromatography module.
- Connect the eluent line from the Sample Injection Valve to the ELUENT IN port of the Dionex DRS 600.
- Connect the ELUENT OUT port of the Dionex DRS 600 to Port 5 of the Injection Valve in the analytical system.