For detailed information on installation of the Dionex DRS 600 on Dionex ICS systems for Neutralization Mode, refer to Thermo Scientific Dionex Application Notes 93 and 94.
NOTE
The Dionex ERS 500e and Dionex AERS 500 Carbonate suppressors are not recommended for use with the Neutralization Mode of Operation. The Dionex DRS 600 should be used.