The Dionex Suppressor Installation Kit (Item # 038018) contains all of the components needed to install and operate a Dionex DRS 600 or Dionex ERS 500e with a pressurized regenerant reservoir. The kit contains:
- Suppressor Installation Parts Kit (Item # 039055)
- A 25-psi regulator (Item # 038201)
- A 4-liter regenerant reservoir (Item # 039164)
To operate a system in the MPIC Suppression Mode, configure the system and use the appropriate sulfuric acid concentration as described in MPIC Suppression Mode Operation.