The Dionex DRS 600 and Dionex AERS 500e can be used with current applied to the suppressor augmented by dilute sulfuric acid in the regenerant solution. If you intend to use an AutoRegen Accessory, configure the system as described in Plumbing for the AutoSuppression External Water Mode, and use the appropriate sulfuric acid concentration instead of water. The Dionex CDRS 600 and Dionex CERS 500e can be used with current applied to the suppressor augmented by diluted boric acid in the regenerant solution.
NOTE
The following installation instructions are based on a system configured with a pressurized water delivery system but apply equally well to peristaltic pump delivery systems.
NOTE
The Dionex AERS 500 Carbonate suppressor is not compatible with the MPIC Suppression Mode of Operation.