The Dionex DRS 600, Dionex ERS 500e and Dionex AERS 500 Carbonate can be used in the Chemical Suppression Mode; however, the total suppression reduced compared to a Chemically Regenerated Suppressor. Thermo Scientific recommends the use of a Chemically Regenerated Suppressor such as the Dionex CRS 500 for chemical suppression applications.