For peristaltic pump plumbing refer to the “MASTERFLEX® C/L® Peristaltic Pump Quick Start Guide” (Item # 065203). For the Dionex DRS 600, Dionex ERS 500e or Dionex AERS 500 Carbonate plumbing, refer to the following figure and pump the Deionized water into the “Regen in” port of the suppressor using the peristaltic pump at the same flow rate as the eluent flow rate.
AutoSuppression External Water Mode plumbing diagram
AutoSuppression External Water Mode plumbing diagram
For installation in the thermal chamber, see Installation in thermal chamber.