System | Installation instructions |
|---|---|
Dionex ICS-3000/5000/5000+/6000 DC | Procedure
|
Dionex Integrion | Procedure
|
Dionex Aquion, Dionex ICS 1100/1600/2100, AS50 thermal chamber TC and CC | Procedure
|
LC30 and LC25 ovens | Procedure
|
NOTE
For best performance and suppressor longevity, the suppressor should be kept as cool as possible; 20° C is ideal.
If using a Dionex DRS 600 in Legacy Mode, a Dionex ERS 500e or Dionex AERS 500 Carbonate, set the required current based on your specific application requirements for column flow rate and temperature in addition to eluent concentration. If using Chromeleon software, the Wizard can greatly assist you in determining the correct requirements.
Refer to Calculate the electrical current for the suppressor for suppressor current calculations and Appendix A for examples.